(S)-(-)-α-(N-t-BOC-Amino)-γ-Butyrolactone

Toimittaja: Apollo Scientific

OR316009-1G OR316009-5G
APOSOR316009-1GEA 21.7 EUR
APOSOR316009-1G APOSOR316009-5G
(S)-(-)-α-(N-t-BOC-Amino)-γ-Butyrolactone
(S)-(-)-α-(N-t-BOC-Amino)-γ-Butyrolactone
Kaava: C₉H₁₅NO₄
Storage Temperature: Huoneenlämpö
MDL numero: MFCD04973958
CAS-numero: 40856-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR