Methyl cyclopentanecarboxylate 97%

Toimittaja: Apollo Scientific

OR310647-5G OR310647-25G
APOSOR310647-5GEA 0 EUR
APOSOR310647-5G APOSOR310647-25G
Methyl cyclopentanecarboxylate 97%
Methyl cyclopentanecarboxylate
Kaava: C₇H₁₂O₂
Kiehumispiste: 158 °C
Tiheys: 0,98 g/cm³ (25 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00019290
CAS-numero: 4630-80-2
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR