(2S,3S,4S,5R)-3,4,5-Tris(benzyloxy)-2-[(benzyloxy)methyl]piperidine hydrochloride

Toimittaja: Apollo Scientific

OR310534-500MG
APOSOR310534-500MGEA 1360 EUR
APOSOR310534-500MG
(2S,3S,4S,5R)-3,4,5-Tris(benzyloxy)-2-[(benzyloxy)methyl]piperidine hydrochloride
(2S,3S,4S,5R)-3,4,5-Tris(benzyloxy)-2-[(benzyloxy)methyl]piperidine hydrochloride
Kaava: C₃₄H₃₇NO₄·ClH
Storage Temperature: Huoneenlämpö
MDL numero: MFCD27957339
CAS-numero: 72983-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR