3-Methyloxetan-3-ol 97%

Toimittaja: Apollo Scientific
Vaara

OR309102-1G OR309102-5G
APOSOR309102-1GEA 66.3 EUR
APOSOR309102-1G APOSOR309102-5G
3-Methyloxetan-3-ol 97%
3-Methyloxetan-3-ol
Kaava: C₄H₈O₂
Storage Temperature: Pakastin
MDL numero: MFCD18909291
CAS-numero: 162816-08-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR