(1S,2R)-2-Amino-cyclohex-3-enecarboxylic acid hydrochloride

Toimittaja: Apollo Scientific

OR307646-1G OR307646-250MG
APOSOR307646-1GEA 1310 EUR
APOSOR307646-1G APOSOR307646-250MG
(1S,2R)-2-Amino-cyclohex-3-enecarboxylic acid hydrochloride
(1S,2R)-2-Amino-cyclohex-3-enecarboxylic acid hydrochloride
Kaava: C₇H₁₁NO₂·ClH
Storage Temperature: Huoneenlämpö
MDL numero: MFCD04112726
CAS-numero: 132487-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR