DL-Phenylglycine ethyl ester hydrochloride 98%

Toimittaja: Apollo Scientific
Varoitus

Synonyms: Ethyl amino(phenyl)acetate hydrochloride, α-(Ethoxycarbonyl)benzylamine hydrochloride, H-DL-2-Phg-OEt·HCl, Ethyl 2-amino-2-phenylacetate hydrochloride

OR26810-1G OR26810-5G OR26810-25G
APOSOR26810-1GEA 103 EUR
APOSOR26810-1G APOSOR26810-5G APOSOR26810-25G
DL-Phenylglycine ethyl ester hydrochloride 98%
DL-Phenylglycine ethyl ester hydrochloride
Kaava: C₁₀H₁₃NO₂·ClH
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00114583
CAS-numero: 879-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR