Pentamethylbenzenesuphonyl chloride, Technical Grade

Toimittaja: Apollo Scientific
Vaara

OR23675-100G OR23675-25G OR23675-5G
APOSOR23675-100GEA 746 EUR
APOSOR23675-100G APOSOR23675-25G APOSOR23675-5G
Pentamethylbenzenesuphonyl chloride, Technical Grade
Pentamethylbenzenesuphonyl chloride
Kaava: C₁₁H₁₅ClO₂S
Sulamispiste: 81…85 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00014722
CAS-numero: 52499-94-2
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR