1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid 97%

Toimittaja: Apollo Scientific
Varoitus

OR2350T-1G OR2350T-5G
APOSOR2350T-1GEA 70.1 EUR
APOSOR2350T-1G APOSOR2350T-5G
1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid 97%
1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid

A highly selective calcium-ion chelating reagent.

Kaava: C₂₂H₂₄N₂O₁₀
Sulamispiste: 189…190 °C
Storage Temperature: Pakastin
MDL numero: MFCD00036255
CAS-numero: 85233-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR