N-Methyldopamine hydrochloride 98%

Toimittaja: Apollo Scientific
Varoitus

OR1575-1G
APOSOR1575-1GEA 304 EUR
APOSOR1575-1G
N-Methyldopamine hydrochloride 98%
N-Methyldopamine hydrochloride
Kaava: C₉H₁₃NO₂·HCl
Sulamispiste: 180…181 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00035074
CAS-numero: 62-32-8
EINECS: 200-528-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR