tert-Butyl-4-{2-[4-(hydrazinocarbonyl)piperidino]ethyl}tetrahydro-1(2H)-pyrazinecarboxylate

Toimittaja: Apollo Scientific

OR14125-1G OR14125-5G
APOSOR14125-1GEA 541 EUR
APOSOR14125-1G APOSOR14125-5G
tert-Butyl-4-{2-[4-(hydrazinocarbonyl)piperidino]ethyl}tetrahydro-1(2H)-pyrazinecarboxylate
tert-Butyl-4-{2-[4-(hydrazinocarbonyl)piperidino]ethyl}tetrahydro-1(2H)-pyrazinecarboxylate
Kaava: C₁₇H₃₃N₅O₃
MDL numero: MFCD06808601
CAS-numero: 874831-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR