1-Cycloheptenylboronic acid 97%

Toimittaja: Apollo Scientific

OR13164-1G OR13164-250MG
APOSOR13164-1GEA 1030 EUR
APOSOR13164-1G APOSOR13164-250MG
1-Cycloheptenylboronic acid 97%
1-Cycloheptenylboronic acid
Kaava: C₇H₁₃BO₂
Storage Temperature: Pakastin
MDL numero: MFCD04115655
CAS-numero: 835882-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR