N-Methylhydroxylammonium chloride 97%

Toimittaja: Apollo Scientific
Varoitus

OR12783-25G OR12783-1G OR12783-5G
APOSOR12783-25GEA 86.2 EUR
APOSOR12783-25G APOSOR12783-1G APOSOR12783-5G
N-Methylhydroxylammonium chloride 97%
N-Methylhydroxylammonium chloride
Kaava: CH₆ClNO
Sulamispiste: 80…86 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012597
CAS-numero: 4229-44-1
EINECS: 224-181-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR