(Methoxymethyl)triphenylphosphonium bromide 98%

Toimittaja: Apollo Scientific
Varoitus

OR10663-1G OR10663-5G
APOSOR10663-1GEA 46.8 EUR
APOSOR10663-1G APOSOR10663-5G
(Methoxymethyl)triphenylphosphonium bromide 98%
(Methoxymethyl)triphenylphosphonium bromide
Kaava: C₂₀H₂₀BrOP
Sulamispiste: 230 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00075442
CAS-numero: 33670-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR