E-Phenylethenylboronic acid 95%

Toimittaja: Apollo Scientific
Varoitus

OR10578-5G OR10578-25G
APOSOR10578-5GEA 70.1 EUR
APOSOR10578-5G APOSOR10578-25G
E-Phenylethenylboronic acid 95%
E-Phenylethenylboronic acid
Kaava: C₈H₉BO₂
Sulamispiste: 146…156 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00963621
CAS-numero: 6783-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR