2-Amino-1,6-dimethyl-1H-imidazo[4,5-b]pyridine

Toimittaja: Apollo Scientific
Varoitus

OR0518T-100MG OR0518T-10MG OR0518T-5MG
APOSOR0518T-100MGEA 2130 EUR
APOSOR0518T-100MG APOSOR0518T-10MG APOSOR0518T-5MG
2-Amino-1,6-dimethyl-1H-imidazo[4,5-b]pyridine
2-Amino-1,6-dimethyl-1H-imidazo[4,5-b]pyridine

A mutagenic cogener of PHIP.

Kaava: C₈H₁₀N₄
Storage Temperature: Pakastin
MDL numero: MFCD00269879
CAS-numero: 132898-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR