2-Thiazolecarbonyl chloride 97%

Toimittaja: Apollo Scientific
Vaara

OR0455-1G OR0455-250MG OR0455-500MG
APOSOR0455-1GEA 211 EUR
APOSOR0455-1G APOSOR0455-250MG APOSOR0455-500MG
2-Thiazolecarbonyl chloride 97%
Kaava: C₄H₂ClNOS
Sulamispiste: 54…58 °C
Storage Temperature: Pakastin
MDL numero: MFCD06658970
CAS-numero: 30216-57-0
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR