2,6-Dibromo-4-isopropylphenyl isocyanate

Toimittaja: Apollo Scientific
Varoitus

OR0333-100MG
APOSOR0333-100MGEA 35.1 EUR
APOSOR0333-100MG
2,6-Dibromo-4-isopropylphenyl isocyanate
2,6-Dibromo-4-isopropylphenyl isocyanate

  • May form small amount of sediment.

Kaava: C₁₀H₉Br₂NO
Storage Temperature: Jääkaappi
CAS-numero: 306935-84-2
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR