Thulium(III) nitrate pentahydrate 99,9%

Toimittaja: Apollo Scientific
Vaara

IN3607-1G IN3607-5G
APOSIN3607-1GEA 86.5 EUR
APOSIN3607-1G APOSIN3607-5G
Thulium(III) nitrate pentahydrate 99,9%
Thulium(III) nitrate pentahydrate
Kaava: N₃O₉Tm·5H₂O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149875
CAS-numero: 36548-87-5
UN: 1477
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR