Europium trinitrate hydrate 99,9%

Toimittaja: Apollo Scientific
Vaara

IN1777-100G IN1777-10G
APOSIN1777-100GEA 1390 EUR
APOSIN1777-100G APOSIN1777-10G
Europium trinitrate hydrate 99,9%
Europium trinitrate hydrate
Kaava: EuN₃O₉·xH₂O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149697
CAS-numero: 100587-95-9
UN: 1477
ADR: 5.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR