Recombinant collagenase H
Toimittaja: Apollo Scientific
BIPR1005-2000C BIPR1005-5000C
APOSBIPR1005-2000CEA 0 EUR
APOSBIPR1005-2000C APOSBIPR1005-5000C
Recombinant collagenase H
Entsyymit
From Clostridium histolyticum and produced in E. Coli and B. Megaterium. Endotoxin negative. No detectable Caseinase activity. Sterile lyophilized powder.