AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride) ≥98%

Toimittaja: Apollo Scientific
Vaara

BIMB2003-1G BIMB2003-500MG BIMB2003-5G
APOSBIMB2003-1GEA 79.2 EUR
APOSBIMB2003-1G APOSBIMB2003-500MG APOSBIMB2003-5G
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride) ≥98%
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)

An irreversible serine protease inhibitor with highwater solubility.


  • Molecular biology grade

Kaava: C₈H₁₁ClFNO₂S
Storage Temperature: Pakastin
MDL numero: MFCD00132962
CAS-numero: 30827-99-7
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR