(3-((3-kolamidopropyyli)dimetyyliammonio)-1-propaanisulfonaatti) ≥99.0%, Ultrapure

Toimittaja: Apollo Scientific
Varoitus

BIMB1085-10G BIMB1085-25G BIMB1085-50G
APOSBIMB1085-10GEA 51.2 EUR
APOSBIMB1085-10G APOSBIMB1085-25G APOSBIMB1085-50G
(3-((3-kolamidopropyyli)dimetyyliammonio)-1-propaanisulfonaatti) ≥99.0%, Ultrapure
(3-((3-kolamidopropyyli)dimetyyliammonio)-1-propaanisulfonaatti)

A nondenaturing zwitterionic detergent for membrane chemistry.


  • Purity (HPLC) >99.0%
  • Purity (TLC) >99.0%
  • Abs. (10% water) @ 260nm <0.1

Kaava: C₃₂H₅₈N₂O₇S
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00081083
CAS-numero: 75621-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR