Stabiliser for proteins and standards, SeramunStab® Stpur
Toimittaja: Apollo Scientific
BIL3118-100ML
APOSBIL3118-100MLEA 87.6 EUR
APOSBIL3118-100ML
Stabiliser for proteins and standards, SeramunStab® Stpur
Proteiinien yhdistämis- ja leimausjärjestelmät
Liquid product colourless to slightly yellow or greenish odourless pH 7,3 to 7,5 boiling point 104 °C. Stabiliser for controls and protein standards.