Motesanib free base 99%

Toimittaja: Apollo Scientific

Synonyms: N-(3,3-Dimethylindolin-6-yl)-2-((pyridin-4-ylmethyl)amino)nicotinamide, AMG706

BIFK0029-10MG BIFK0029-50MG BIFK0029-100MG
APOSBIFK0029-10MGEA 65.8 EUR
APOSBIFK0029-10MG APOSBIFK0029-50MG APOSBIFK0029-100MG
Motesanib free base 99%
Motesanib
Kaava: C₂₂H₂₃N₅O
Sulamispiste: 140…151 °C
Storage Temperature: Pakastin
MDL numero: MFCD10567689
CAS-numero: 453562-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR