Imatinib mesylate ≥99%

Toimittaja: Apollo Scientific

BIFK0026-1G BIFK0026-5G BIFK0026-250MG
APOSBIFK0026-1GEA 117 EUR
APOSBIFK0026-1G APOSBIFK0026-5G APOSBIFK0026-250MG
Imatinib mesylate ≥99%
Imatinib mesylate

Imatinib is the active ingredient in the drug sold under the trade name Gleevec® in USA or Glivec in Europe and Australia.

Kaava: C₂₉H₃₁N₇O·CH₄SO₃
Sulamispiste: 203…224 °C
Storage Temperature: Pakastin
MDL numero: MFCD04307699
CAS-numero: 220127-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR