Fluorescein diacetate

Toimittaja: Apollo Scientific
Varoitus

BIF4105-25G BIF4105-5G
APOSBIF4105-25GEA 132 EUR
APOSBIF4105-25G APOSBIF4105-5G
Fluorescein diacetate
Fluorescein diacetate
Kaava: C₂₄H₁₆O₇
Sulamispiste: 200…203 °C
Storage Temperature: Pakastin
MDL numero: MFCD00005062
CAS-numero: 596-09-8
EINECS: 209-877-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR