EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride)

Toimittaja: Apollo Scientific
Vaara

BIE1308-100G BIE1308-500G BIE1308-1KG
APOSBIE1308-100GEA 26.3 EUR
APOSBIE1308-100G APOSBIE1308-500G APOSBIE1308-1KG
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride)
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride)

EDAC is a water soluble condensing reagent.

Kaava: C₈H₁₇N₃·HCl
Storage Temperature: Pakastin
MDL numero: MFCD00012503
CAS-numero: 25952-53-8
EINECS: 247-361-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR