2,3,4-Tri-O-acetyl-α-L-arabinopyranosyl bromide ≥98%

Toimittaja: Apollo Scientific

BICL4259-1G BICL4259-2G BICL4259-5G
APOSBICL4259-1GEA 395 EUR
APOSBICL4259-1G APOSBICL4259-2G APOSBICL4259-5G
2,3,4-Tri-O-acetyl-α-L-arabinopyranosyl bromide ≥98%
2,3,4-Tri-O-acetyl-α-L-arabinopyranosyl bromide
Kaava: C₁₁H₁₅BrO₇
Storage Temperature: Pakastin
MDL numero: MFCD00057497
CAS-numero: 75247-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR