1-O-Galloyl β-D-glucopyranoside ≥98%

Toimittaja: Apollo Scientific
Varoitus

BICL4108-250MG BICL4108-50MG BICL4108-100MG
APOSBICL4108-250MGEA 1390 EUR
APOSBICL4108-250MG APOSBICL4108-50MG APOSBICL4108-100MG
1-O-Galloyl β-D-glucopyranoside ≥98%
1-O-Galloyl β-D-glucopyranoside
Kaava: C₁₃H₁₆O₁₀
Storage Temperature: Pakastin
MDL numero: MFCD11975493
CAS-numero: 13405-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR