N-Butyldeoxynojirimycin hydrochloride 99% min

Toimittaja: Apollo Scientific

BICL4039-10MG BICL4039-25MG BICL4039-5MG
APOSBICL4039-10MGEA 408 EUR
APOSBICL4039-10MG APOSBICL4039-25MG APOSBICL4039-5MG
N-Butyldeoxynojirimycin hydrochloride 99% min
N-Butyldeoxynojirimycin hydrochloride
Kaava: C₁₀H₂₁NO₄·HCl
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00269940
CAS-numero: 210110-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR