SPDP (N-Succinimidyl-3-(2-pyridyldithio)propionate)

Toimittaja: Apollo Scientific
Varoitus

BICL210-25MG
APOSBICL210-25MGEA 103 EUR
APOSBICL210-25MG
SPDP (N-Succinimidyl-3-(2-pyridyldithio)propionate)
SPDP (N-Succinimidyl-3-(2-pyridyldithio)propionate)

Primary amine & Sulphhydryl reactiveThiolation -CleavableRef: Cumber,J.Aet al(1985)Methods Enzymol.112, 207-224

Kaava: C₁₂H₁₂N₂O₄S₂
Sulamispiste: 84…86 °C
Storage Temperature: Pakastin
MDL numero: MFCD00009636
CAS-numero: 68181-17-9
EINECS: 269-034-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR