H-L-Dap(boc)-OMe·HCl

Toimittaja: Apollo Scientific

BIBA1021-1G BIBA1021-5G BIBA1021-10G
APOSBIBA1021-1GEA 103 EUR
APOSBIBA1021-1G APOSBIBA1021-5G APOSBIBA1021-10G
H-L-Dap(boc)-OMe·HCl
H-L-Dap(boc)-OMe·HCl
Kaava: C₉H₁₉ClN₂O₄
Storage Temperature: Jääkaappi
MDL numero: MFCD00798684
CAS-numero: 114559-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR