4-Nitrophenyl phosphate, bis(cyclohexylammonium) salt

Toimittaja: Apollo Scientific

BIB6134-10G BIB6134-50G BIB6134-5G
APOSBIB6134-10GEA 285 EUR
APOSBIB6134-10G APOSBIB6134-50G APOSBIB6134-5G
4-Nitrophenyl phosphate, bis(cyclohexylammonium) salt
4-Nitrophenyl phosphate, bis(cyclohexylammonium) salt
Kaava: C₆H₆NO₆P·2C₆H₁₃N
Storage Temperature: Pakastin
MDL numero: MFCD00013173
CAS-numero: 52483-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR