L-Dehydroascorbic acid

Toimittaja: Apollo Scientific
Varoitus

BIB6039-1G BIB6039-5G BIB6039-10G
APOSBIB6039-1GEA 87.6 EUR
APOSBIB6039-1G APOSBIB6039-5G APOSBIB6039-10G
L-Dehydroascorbic acid
L-Dehydroascorbic acid
Kaava: C₆H₆O₆
Sulamispiste: 228 °C
Storage Temperature: Pakastin
MDL numero: MFCD00066467
CAS-numero: 490-83-5
EINECS: 207-720-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR