L-Leucine(4-methyl-7-coumarinylamide) hydrochloride

Toimittaja: Apollo Scientific
Varoitus

Synonyms: H-Leu-AMC·HCl

BIB1441-100MG BIB1441-250MG
APOSBIB1441-100MGEA 73 EUR
APOSBIB1441-100MG APOSBIB1441-250MG
L-Leucine(4-methyl-7-coumarinylamide) hydrochloride
L-Leucine(4-methyl-7-coumarinylamide) hydrochloride
Kaava: C₁₆H₂₀N₂O₃·HCl
Sulamispiste: 283 °C
Storage Temperature: Pakastin
MDL numero: MFCD00043262
CAS-numero: 62480-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR