2-Naphthyl-ß-D-galactopyranoside

Toimittaja: Apollo Scientific
Varoitus

BIB1427-250MG BIB1427-500MG
APOSBIB1427-250MGEA 106 EUR
APOSBIB1427-250MG APOSBIB1427-500MG
2-Naphthyl-ß-D-galactopyranoside
2-Naphthyl-ß-D-galactopyranoside
Kaava: C₁₆H₁₈O₆
Storage Temperature: Pakastin
MDL numero: MFCD00004056
CAS-numero: 33993-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR