7-[4-(2,2-Diphosphonoethyl)piperazin-1-yl]-1-ethyl-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid dihydrobromide

Toimittaja: Apollo Scientific

BIA0220-1G BIA0220-5G
APOSBIA0220-1GEA 0 EUR
APOSBIA0220-1G APOSBIA0220-5G
7-[4-(2,2-Diphosphonoethyl)piperazin-1-yl]-1-ethyl-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid dihydrobromide
7-[4-(2,2-Diphosphonoethyl)piperazin-1-yl]-1-ethyl-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid dihydrobromide
Kaava: C₁₇H₂₃FN₄O₉P₂·2BrH
Storage Temperature: Huoneenlämpö
MDL numero: MFCD18072177
CAS-numero: 434341-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR