Phleomycin

Toimittaja: Apollo Scientific
Varoitus

BI3852-100MG BI3852-50MG BI3852-25MG
APOSBI3852-100MGEA 308 EUR
APOSBI3852-100MG APOSBI3852-50MG APOSBI3852-25MG
Phleomycin
Phleomycin

Phleomycin is a glycopeptide antibiotic of the bleomycin family that exhibits in vivo activity against bacteria, eukaryotic microorganisms, plant and animal cells.

Storage Temperature: Pakastin
MDL numero: MFCD00131846
CAS-numero: 11006-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR