Anti-ALDH3A1 Rabbit Polyclonal Antibody

Toimittaja: ANTIBODIES.COM
A98931-100
ANTIA98931-100EA 475 EUR
ANTIA98931-100
Anti-ALDH3A1 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody to ALDH3A1 for WB and ELISA with samples derived from human and rat.

  • Validated applications: WB, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: ALDH3A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR