Anti-RHOD Rabbit Polyclonal Antibody

Toimittaja: ANTIBODIES.COM
A9879-100
ANTIA9879-100EA 415 EUR
ANTIA9879-100
Anti-RHOD Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody to RHOD for WB, IHC and ICC/IF with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, ICC/IF
  • Recommended dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:20

Type: Primary
Antigen: RHOD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR