Anti-Protein Kinase D2 Rabbit Polyclonal Antibody

Toimittaja: ANTIBODIES.COM
A94022-100
ANTIA94022-100EA 475 EUR
ANTIA94022-100
Anti-Protein Kinase D2 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody to PKD2 (phospho Ser876) for WB and ELISA with samples derived from Human, Mouse and Rat.

  • Validated applications: WB, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:60000

Type: Primary
Antigen: Protein Kinase D2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR