Anti-APOBEC2 Goat Polyclonal Antibody

Toimittaja: ANTIBODIES.COM
A84474-100
ANTIA84474-100EA 475 EUR
ANTIA84474-100
Anti-APOBEC2 Goat Polyclonal Antibody
Vasta-aineet
Goat polyclonal antibody to APOBEC2 for ELISA and WB with samples derived from human.

  • Validated applications: ELISA, W

Type: Primary
Antigen: APOBEC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR