Anti-SLC12A4 Goat Polyclonal Antibody

Toimittaja: ANTIBODIES.COM
A84410-100
ANTIA84410-100EA 475 EUR
ANTIA84410-100
Anti-SLC12A4 Goat Polyclonal Antibody
Vasta-aineet
Goat polyclonal antibody to SLC12A4 for ELISA, WB and IHC with samples derived from human.

  • Validated applications: ELISA, WB, IH

Type: Primary
Antigen: SLC12A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR