Anti-Lime1 Goat Polyclonal Antibody

Toimittaja: ANTIBODIES.COM
A84030-100
ANTIA84030-100EA 475 EUR
ANTIA84030-100
Anti-Lime1 Goat Polyclonal Antibody
Vasta-aineet
Goat polyclonal antibody to LIME1 for ELISA and WB with samples derived from mouse.

  • Validated applications: ELISA, W

Type: Primary
Antigen: Lime1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
New Product
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR