Human TENR ELISA Kit

Toimittaja: ANTIBODIES.COM

A76101-96
ANTIA76101-96EA 600 EUR
ANTIA76101-96
Human TENR ELISA Kit
Bio-assayt ELISA-kitit

Human TENR ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human TENR in serum, plasma, tissue homogenates, and other biological fluids.


  • Ready to use ELISA kit
  • Detection range: 0,313 to 20 ng/ml
  • Sensitivity: 0,188 ng/ml
  • Sample type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%


Human TENR ELISA kit (A76101) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human TENR in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for TENR has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the TENR present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-TENR Antibody, which binds the captured TENR present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of TENR captured in each well. The concentration of TENR can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Tilaustiedot: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

New Product

Order Now

TEKNISET TIEDOT

Environmentally Preferable
Target protein TENR
Primary antibody reactivity Human
Määritystyyppi Sandwich
Määrityksen periaate Quantitative
Isäntä
Muoto Pre-coated
Määrityksen kesto Multiple steps
Herkkyys 0,188 ng/ml
Havaintoalue 0,313 - 20 ng/ml
Havaitsemismenetelmä Colorimetric
Ristireaktiivisuus Human TENR ELISA Kit exhibits high specificity and excellent specificity for the detection of human TENR. No significant cross-reactivity or interference between TENR and analogues was observed.
Näytteen tyyppi Serum, plasma, tissue homogenates and other biological fluids
Näytteen tilavuus 100 μl
Time to Results 4 h 30 min
Regulatiivinen status RUO
Säilytyslämpötila 4 °C
Säilyvyys Store for 6 months at 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR