Anti-IL-3 Mouse Monoclonal Antibody [clone: IL3/4003]

Toimittaja: ANTIBODIES.COM
A315176-100
ANTIA315176-100EA 680 EUR
ANTIA315176-100
Anti-IL-3 Mouse Monoclonal Antibody [clone: IL3/4003]
Vasta-aineet
Mouse monoclonal [IL3/4003] antibody to IL-3 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ml

Type: Primary
Antigen: IL-3
Clonality: Monoclonal
Clone: IL3/4003
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR