Mouse FSTL1/FRP ELISA Kit

Toimittaja: ANTIBODIES.COM

A312756-96
ANTIA312756-96EA 545 EUR
ANTIA312756-96
Mouse FSTL1/FRP ELISA Kit
Bio-assayt ELISA-kitit

Mouse FSTL1/FRP ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse FSTL1/FRP in serum, plasma, and other biological fluids.


  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection range: 0,75 to 12 ng/ml
  • Sensitivity: 0,024 ng/ml
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra-assay: CV <8%, Inter-assay: CV <10%


Mouse FSTL1/FRP ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse FSTL1/FRP in serum, plasma or other biological fluids. An antibody specific for FSTL1/FRP has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the FSTL1/FRP present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-FSTL1/FRP antibody, which also binds the FSTL1/FRP present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-FSTL1/FRP antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-FSTL1/FRP antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of FSTL1/FRP captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of FSTL1/FRP in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Tilaustiedot: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR