Human Plexin B1 ELISA Kit

Toimittaja: Antibodies.com

A311086-96
ANTIA311086-96EA 545 EUR
ANTIA311086-96
Human Plexin B1 ELISA Kit
Bio-assayt ELISA-kitit

Human Plexin B1 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Plexin B1 in serum, plasma, and other biological fluids.


  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 400 to 6400 ng/L
  • Sensitivity: 25,84 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%


Human Plexin B1 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Plexin B1 in serum, plasma or other biological fluids. An antibody specific for Plexin B1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Plexin B1 present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-Plexin B1 antibody, which also binds the Plexin B1 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Plexin B1 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Plexin B1 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of Plexin B1 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Plexin B1 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.


Tilaustiedot: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR