Anti-Calprotectin Mouse Monoclonal Antibody [Clone: S100A9/1011]

Toimittaja: ANTIBODIES.COM
A254334-100
ANTIA254334-100EA 520 EUR
ANTIA254334-100
Anti-Calprotectin Mouse Monoclonal Antibody [Clone: S100A9/1011]
Vasta-aineet
Mouse monoclonal [S100A9/1011] antibody to Calprotectin for IHC with samples derived from human.

  • Validated Applications: IHC
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Calprotectin
Clonality: Monoclonal
Clone: S100A9/1011
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR