Anti-Cyclin D1 Mouse Recombinant Antibody [clone: rCCND1/4752]

Toimittaja: ANTIBODIES.COM
A253035-100
ANTIA253035-100EA 520 EUR
ANTIA253035-100
Anti-Cyclin D1 Mouse Recombinant Antibody [clone: rCCND1/4752]
Vasta-aineet
Recombinant mouse monoclonal [rCCND1/4752] antibody to Cyclin D1 for IHC-P with samples derived from Human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Cyclin D1
Clonality: Recombinant
Clone: rCCND1/4752
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR